EMAIL THIS PAGE TO A FRIEND
Preparative biochemistry & biotechnology
Synthesis and analgesic activity of N,N'-dicarbonyltryptamines.
PMID 10919562
Abstract
5-Methoxytryptamine and L-tryptophan methyl ester were acylated with malonic acid, dimethyl malonate, or succinic anhydride to produce the corresponding N,N'-dicarbonyltryptamine derivatives. The analgesic activity was evaluated by the tail flick test. All of the compounds exhibited desirable analgesic potency. This result is consistent with that of N-(N-acetyl-L-tryptophanyl)-5-methoxytryptamine and confirmed that introducing substituted tryptamine into the amide chain of melatonin does enhance analgesic potency.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
136441
Dimethyl malonate, 98%
C5H8O4
63380
Dimethyl malonate, purum, ≥96.0% (GC)
C5H8O4