EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
Polysaccharide labeling with N-methylisatoic anhydride: generation of ultraviolet chromophores and blue fluorophores.
PMID 10933873
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
129887
N-Methylisatoic anhydride, technical, 90%
C9H7NO3