EMAIL THIS PAGE TO A FRIEND
Steroids
Selective activation of liver X receptor alpha by 6alpha-hydroxy bile acids and analogs.
PMID 10936612
Abstract
We have found that certain natural 6alpha-hydroxylated bile acids are receptor-specific activators of nuclear liver X receptor alpha (LXRalpha) (NR1H3), a nuclear receptor regulating the expression of the cholesterol 7alpha-hydroxylase gene, coding for the rate-limiting enzyme in the major pathway of bile acid synthesis. The LXR homolog, ubiquitous nuclear receptor (UR/LXRbeta) (NR1H2), was also activated by these bile acids, but at higher concentrations than for LXRalpha. Synthetic 6alpha-hydroxylated bile acid analogs were synthesized with LXRalpha-selective agonistic activity, with potential to modulate cholesterol catabolism in hypercholesterolemia.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T0682
Sodium taurohyodeoxycholate hydrate, ≥98%
C26H44NNaO6S · xH2O