EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Metalloporphyrin catalyzed oxidation of N-hydroxyguanidines: a biomimetic model for the H2O2-dependent activity of nitric oxide synthase.
PMID 10937746
Abstract
A chemical model for the H2O2 promoted oxidation by nitric oxide synthase (NOS) has been developed. Biomimetic oxidations were carried out using H2O2 and tetrakis(perfluorophenyl)porphyrinato-iron(III) chloride (FeTPPF20) as a catalyst. Similarly to NOS our model system produces Ndelta-cyanoornithine, citrulline and NO from NOHA and did not oxidize arginine itself. Based on these results we propose a peroxide shunt to be involved in the catalytic cycle of NOS. To the best of our knowledge this is the first chemical system that semiquantitatively mimics NOS activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H7654
Hydroxyguanidine sulfate salt
CH5N3O · 0.5H2O4S