EMAIL THIS PAGE TO A FRIEND
Blood
Human erythrocyte pyrimidine 5-nucleotidase, PN-I, is identical to p36, a protein associated to lupus inclusion formation in response to alpha-interferon.
PMID 10942414
Abstract
Erythrocyte maturation is accompanied by RNA degradation and release of mononucleotides. We have previously purified PN-I, a pyrimidine nucleotidase whose deficiency is associated with hemolytic anemia. Computer-aided analysis of PN-I tryptic and CNBr peptide sequences revealed substantial identity with tryptic peptide sequences reported for p36, an alpha-interferon-induced protein. PN-I partial sequences were matched through the expressed sequence tag database with different human complementary DNA (cDNA) clones, whose sequences were exploited to screen a human placenta cDNA library. PN-I cDNA, coding for a 286-residue protein, was expressed in Escherichia coli, yielding a fully active recombinant enzyme. The recombinant protein sequence comprised the peptide sequences determined for PN-I and p36. Rabbit antisera raised against two peptides deriving from p36 and PN-I tryptic digestions, respectively, recognized both wild-type and recombinant PN-I. Molecular properties of the two proteins were essentially the same. We conclude that p36 and PN-I are identical proteins. (Blood. 2000;96:1596-1598)