EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
New methods for determination of glyphosate and (aminomethyl)phosphonic acid in water and soil.
PMID 10950288
Abstract
New methods were developed to determine glyphosate, N-(phosphonomethyl)glycine, and its major metabolite, (aminomethyl)phosphonic acid in groundwater and soil. The methods involve ligand-exchange, anion-exchange and derivatisation and final identification and quantification by GC-MS. The limits of quantification in this experiment were 0.1 microg l(-1) for both compounds in water and 0.006 microg g(-1) for both compounds in soil. Decomposition in soil and occurrence in groundwater of the herbicide glyphosate was studied after its application for weed control on a Swedish railway embankment.