EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
Chemiluminescence derivatization of methylglyoxal using 2-aminonicotinic acid.
PMID 10959598
Abstract
To develop a sensitive and selective chemiluminometric method for the determination of methylglyoxal, we used 2-aminonicotinic acid as the chemiluminescence derivatization reagent. 2-Aminonicotinic acid reacts with methylglyoxal in an acidic solution at 37 degrees C for 4 h and gave a chemiluminescence in N,N'-dimethylformamide containing sodium tert-butoxide. The detection limit (blank intensity plus three-times its standard deviation of methylglyoxal) for methylglyoxal is 233 fmol in the reaction mixture.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A68300
2-Aminopyridine-3-carboxylic acid, 98%
C6H6N2O2