EMAIL THIS PAGE TO A FRIEND
Current opinion in chemical biology
Bradykinin antagonists: new opportunities.
PMID 10959767
Abstract
The pro-inflammatory, pain producing, and cardiovascular effects of bradykinin B2 receptor activation are well characterized. Bradykinin B1 receptors also produce inflammation and pain. Therefore, antagonists are expected to be anti-inflammatory/analgesic drugs. Other exploitable clinical opportunities may exist. The newly discovered non-peptide B2 receptor antagonists and the equivalent B1 receptor pharmacological agents, which are in the pipeline, are suitable preclinical tools to properly evaluate potential utilities.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H157
HOE 140, ≥94%
C59H89N19O13S