EMAIL THIS PAGE TO A FRIEND
Cytogenetics and cell genetics
Assignment of the mitochondrial translation elongation factor Ts gene (TSFM) to human chromosome 12 bands q13-->q14 by in situ hybridization and with somatic cell hybrids.
PMID 10965106