EMAIL THIS PAGE TO A FRIEND
Applied and environmental microbiology
Lactococcus lactis as a cell factory for high-level diacetyl production.
PMID 10966436
Abstract
We report the engineering of Lactococcus lactis for the efficient conversion of sugar into diacetyl by combining NADH-oxidase overproduction and alpha-acetolactate decarboxylase inactivation. Eighty percent of the carbon flux was found to be rerouted via alpha-acetolactate to the production of diacetyl by preloading the cells with NADH-oxidase before their use as a cell factory.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
00878
(±)-2-Acetoxypropionic acid, ≥97.0% (GC)
C5H8O4