EMAIL THIS PAGE TO A FRIEND
Current microbiology
Identification of heptadecanoic and C19 cyclopropane fatty acids in the lipid fraction of Zymomonas mobilis.
PMID 10977901
Abstract
In the present paper the presence of heptadecanoic acid and C19 cyclopropane fatty acid in Zymomonas mobilis is reported. The former has not been mentioned as a component of the lipid fraction of this microorganism, and the presence of the latter is not well documented.