EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Solid-phase synthesis of acridine-based threading intercalator peptides.
PMID 10987431
Abstract
The preparation of a novel acridine-based amino acid is reported. This N-Alloc-protected monomer can be coupled and deprotected under solid-phase peptide synthesis procedures to create acridine peptide conjugates as potential threading intercalators. A peptide containing this novel amino acid undergoes spectral changes in the presence of duplex DNA and RNA consistent with intercalative binding.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
368466
4-Aminobenzylamine, 99%
C7H10N2