EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Preparation of aryl isothiocyanates via protected phenylthiocarbamates and application to the synthesis of caffeic acid (4-isothiocyanato)phenyl ester.
PMID 10987967
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
476439
2,6-Dimethylphenyl isothiocyanate, 98%
C9H9NS
496642
3-Cyanophenyl isothiocyanate, 98%
C8H4N2S