EMAIL THIS PAGE TO A FRIEND
Organic letters
Use of thallium(I) ethoxide in Suzuki cross coupling reactions.
PMID 10990429
Abstract
[reaction: see text]Thallium(I) ethoxide promotes Suzuki cross couplings for a range of vinyl- and arylboronic acids with vinyl and aryl halide partners in good to excellent yields. This reagent offers distinct advantages over thallium(I) hydroxide in terms of commercial availability, stability, and ease of use.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
149845
Thallium(I) ethoxide
C2H5OTl