EMAIL THIS PAGE TO A FRIEND
Voprosy kurortologii, fizioterapii, i lechebnoi fizicheskoi kultury
[The use of physical factors in the rehabilitative treatment of patients with stone fragments in the upper urinary tract after extracorporeal shockwave lithotripsy].
PMID 11008569
Abstract
155 patients with fragments of the crushed stones in the upper urinary tract after lithotripsy were divided into 3 groups. 60 patients of group 1 received dynamic amplipulse therapy, oral mineral water, iodobromine baths. Elimination of the fragments was observed in 91.7% of the patients. Group 2 patients (n = 60) were exposed to local vibration, ultrasound and iodobromine baths. The effect occurred in 96.7% of the cases. 35 patients of group 3 were exposed to impulse low-frequency magnetic field and took iodobromine baths. The elimination of the fragments occurred in 62.9% of the cases after a single treatment course.
Related Materials