EMAIL THIS PAGE TO A FRIEND
Organic letters
Facile, completely regioselective 1,4-hydrogenations of C(60)-diaryltetrazine monoadducts
PMID 11009353
Abstract
Thermal Diels-Alder reactions between C(60) and electron-deficient 3, 6-diaryl-1,2,4,5-tetrazines yield monoadducts possessing a diaryldihydropyridazine function nested atop the fullerene. The diaryldihydropyridazine functions direct a completely regioselective 1,4-hydrogenation, resulting in a racemic mixture of bifunctional bisadduct (+/-)-3.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
403555
3,6-Diphenyl-1,2,4,5-tetrazine, 98%
C14H10N4