EMAIL THIS PAGE TO A FRIEND
Organic letters
Efficient asymmetric synthesis of radicicol dimethyl ether: a novel application of ring-forming olefin metathesis.
PMID 11009362
Abstract
A concise, stereospecific synthesis of radicicol dimethyl ether is presented. The strategy relies on a convergent three-stage assembly of the 14-membered lactone which has, as a key transformation, a novel ring-forming metathesis reaction utilizing a vinyl epoxide.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
54957
Methyl (R)-3-hydroxybutyrate, puriss., ≥99.0% (sum of enantiomers, GC)
C5H10O3