EMAIL THIS PAGE TO A FRIEND
American journal of physiology. Heart and circulatory physiology
Interactions of hemoglobin with hydrogen peroxide alters thiol levels and course of endothelial cell death.
PMID 11009476
Abstract
We investigated cellular injury and death induced by ultrapure human Hb (HbA(0)) and its diaspirin cross-linked derivative DBBF-Hb in normal and glutathione (GSH)-depleted bovine aortic endothelial cells subjected to hydrogen peroxide (H(2)O(2)). HbA(0) underwent extensive degradation and heme loss, whereas DBBF-Hb persisted longer in its ferryl (Fe(4+)) form. The formation of ferryl HbA(0) or ferryl DBBF-Hb was associated with a significant decrease in endothelial cell GSH compared with the addition of H(2)O(2) or Hbs alone. This effect was inhibited by catalase, but not by superoxide dismutase or deferoxamine mesylate. The presence of HbA(0) and DBBF-Hb reduced H(2)O(2)-induced apoptosis, as measured by cell morphology, annexin V binding assay, and caspase inhibition, consistent with the ability to consume H(2)O(2) in an enzyme-like fashion. However, the pattern of cell death and injury produced by HbA(0) and DBBF-Hb appeared to be distinctly different among proteins as well as among cells with and without GSH. These findings may have important implications for the use of cell-free Hb as oxygen therapeutics in patients with coexisting pathologies who may lack antioxidant protective mechanisms.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H0267 Hemoglobin A0, Ferrous Stabilized human, lyophilized powder