EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Contact allergy due to propineb.
PMID 11016679
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45643
Propineb, PESTANAL®, analytical standard