EMAIL THIS PAGE TO A FRIEND
Nihon rinsho. Japanese journal of clinical medicine
[Leuprorelin acetate depot].
PMID 11022712
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L0376000 Leuprorelin, European Pharmacopoeia (EP) Reference Standard
C59H84N16O12