EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Selective boron-containing thrombin inhibitors--X-ray analysis reveals surprising binding mode.
PMID 11026541
Abstract
Based on the structural comparison of the S1 pocket in different trypsin-like serine proteases, a series of Boc-D-trimethylsilylalanine-proline-boro-X pinanediol derivatives, with boro-X being different amino boronic acids, have been synthesized as inhibitors of thrombin. Among the novel compounds, a number of derivatives were synthesized which appeared to have side-chain variants too big to fit into the S1 pocket. Nevertheless, these compounds inhibited thrombin in the nM range. The X-ray structure of one of these inhibitors bound to the active side of thrombin reveals that a new binding mode is responsible for these surprising results.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
80593
(1R)-(+)-cis-Pinane, analytical standard
C10H18