EMAIL THIS PAGE TO A FRIEND
The Cochrane database of systematic reviews
Penicillamine for treating rheumatoid arthritis.
PMID 11034719
Abstract
To estimate the short-term effects of D-penicillamine for the treatment of rheumatoid arthritis (RA). We searched the Cochrane Musculoskeletal Group's trials register, the Cochrane Controlled Trials Register (issue 3, 2000) and Medline up to and including August 2000 and Embase from 1988-2000. We also carried out a handsearch of the reference lists of the trials retrieved from the electronic search. All randomized controlled trials and controlled clinical trials comparing D-penicillamine against placebo in patients with rheumatoid arthritis. The methodological quality of the trials was assessed independently by two reviewers (CS, EB) and checked by a third (MS) using a validated quality assessment tool (Jadad 1996). Rheumatoid arthritis outcome measures were extracted from the publications for the six-month endpoint and stratified according to D-penicillamine dosages: low (<500mg/day), moderate (500 to <1000mg/day) and high (1000 mg/day or greater). Data was abstracted by one reviewer and checked by a second (CS, MS). The pooled analysis was performed using the standardized mean difference for joint counts, pain and global assessments. The weighted mean difference was used for erythrocyte sedimentation rate (ESR). Toxicity was evaluated with pooled odds ratios for withdrawals and adverse reactions. A chi-square test was used to assess heterogeneity among trials. Fixed effects models were used throughout, since no statistical heterogeneity was found. Six trials were identified, with 425 patients randomized to D-penacillamine and 258 to placebo. A statistically significant benefit was observed for D-penicillamine when compared to placebo for all three-dose ranges and for most outcome measures including: tender joint counts, pain, physician's global assessments and ESR. The standardized weighted mean differences between treatment and placebo in moderate doses were -0.51 [95% CI -0.88, -0.14] for tender joint counts, -0.56 (95% CI -0.87, -0.26) for pain and -0.97 (95% CI -1.25, -0.70) for global assessment. The difference for ESR was -10.6 mm/hr. Similar results were observed for the higher dose group. Total withdrawals were significantly higher in the moderate and high dosage D-penicillamine groups (OR=1.63 and 2.13 respectively), mostly due to increased adverse reactions (OR = 2.60 and 4.95 respectively), including renal and hematological abnormalities. D-penicillamine appears to have a clinically and statistically significant benefit on the disease activity of patients with rheumatoid arthritis. Its efficacy appears to be similar to that of other disease modifying anti-rheumatic drugs (DMARDs), but with a significantly higher toxicity. Its effects on long-term functional status and radiological progression are not clear from this review.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P0310000
Penicillamine, European Pharmacopoeia (EP) Reference Standard
C5H11NO2S