EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica
[Determination of eugenol, methyleugenol and methylisoeugenol in volatile oil of differently processed nutmeg].
PMID 11038913
Abstract
A HPLC method for the determination of eugenol, methyleugenol and methylisoeugenol in the volatile oil of differently processed natmeg has been used. The result has shown that the content of eugenol is only sliahtly changed before and after processing, but that of methyleugenol and methylisoeugenol has obviously increased.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D136905
1,2-Dimethoxy-4-propenylbenzene, 99%
C11H14O2
W247618
Methyl isoeugenol, ≥98%, FG
C11H14O2