EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica
[GC-MS analysis of constituents of essential oils from stems of Ephedra sinica Stapf, E. intermedia Schrenk et C.A. Mey. and E. equisetina Bge].
PMID 11038918
Abstract
The essential oils from the dried stems of Ephedra sinica, E. intermedia and E. equisetina were analyzed by GC-MS qualitatively and GC quantitatively. One hundred and twenty-seven constituents were identified, l-alpha-terpineol (31.64%) in E. sinica, 1,4-cineole (12.80%) in E. intermedia and hexadecanoic acid (26.22%) in E. equisetina being the main constituents.
Related Materials