EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica
[Comparative study on nutmeg, mace and their processed products].
PMID 11038942
Abstract
The contents of volatile oil, myristicin, safrol and methyleugenol, as well as the composition and content of fatty oil in nutmeg, mace and their processed products were comparatively studied. The composition of volatile oil and the detection of trimyristin were compared by TLC. A scientific basis for the development and application of mace has thus been provided.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T5141
Glyceryl trimyristate, ≥99%
C45H86O6