EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Configurational analysis of chiral acids as O-trifluoroacetylated (-)-menthyl esters by achiral dual-capillary column gas chromatography.
PMID 11043786
Abstract
The simultaneous enantiomeric separation of 30 racemic acids including 24 hydroxy acids in a single analysis is described for the determination of their absolute configurations. It involves the conversion of each enantiomer into diastereomeric O-trifluoroacetylated (-)-menthyl ester for the direct separation by gas chromatography on achiral dual-capillary columns of different polarities, with subsequent identification and chiral discrimination by retention index (I) library matching. Among the acids studied, the enantiomers of 27 acids were discriminatively resolved on both non-polar DB-5 and the intermediate-polar DB-17 columns with resolution factors in the range of 0.7-7.7 and separation factors in the range of 1.002-1.021. Enantiomers of 3-hydroxybutyric and alpha-methoxyphenylacetic acids were partially resolved on DB-5 (resolution factor of 0.9), but not resolved on DB-17, while the baseline resolution for 3-hydroxydecanoic acid and the minimal separation on the peak top (resolution factor less than 0.7) for 2-hydroxyglutaric acid were achieved on DB-17 but not on DB-5. The temperature-programmed I values measured on both columns were characteristic of each enantiomer and thus simple I matching with the reference values was useful in cross-checking for their chemical identification and the chiral discrimination as well. When applied to a clinical urine sample, the present method allowed positive identification of endogenous (S)-lactic acid and (S)-2-hydroxybutyric acid along with (R)-3-hydroxybutyric acid.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
55397
(R)-3-Hydroxybutyric acid, analytical standard
C4H8O3
116807
3-Phenylbutyric acid, 98%
C10H12O2