EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
Hydroxylation of nitrated naphthalenes with KO2/crown ether.
PMID 11045464
Abstract
Superoxide radical anion (O2*-), generated by KO2/crown ether, is effective for hydroxylation of nitronaphthalenes. When mono- and di-nitronaphthalenes are treated with KO2/crown ehter, hydroxylation results at the electron-deficient site caused by the electron withdrawing effect of the substituted nitro group. Kinetic experiments suggest that the hydroxylation proceeds by two different mechanisms dependent on the first one-electron reduction potential of nitronaphthalenes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
310492
Potassium dioxide, chunks, 5-10 mm
KO2
278904
Potassium dioxide, powder
KO2