EMAIL THIS PAGE TO A FRIEND
Methods in molecular biology (Clifton, N.J.)
Amino acid analysis by high-performance liquid chromatography after derivatization with 1-fluoro-2,4-dinitrophenyl-5-L-alanine amide (Marfey's reagent).
PMID 11050716