EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Rapid and sensitive determination of 4-nitrophenol, 3-methyl-4-nitrophenol, 4,6-dinitro-o-cresol, parathion-methyl, fenitrothion, and parathion-ethyl by liquid chromatography with electrochemical detection.
PMID 11052691
Abstract
Liquid chromatography with electrochemical detection has been used to determine various nitropesticides, DNOC, fenitrothion, and parathion (methyl and ethyl), and some of their main metabolites, 4-nitrophenol for parathion (methyl and ethyl) and 3-methyl-4-nitrophenol for fenitrothion, by using indirect detection. Analysis of them in river water samples has been performed without a preconcentration step. The recovery efficiencies of the tested compounds yielded values between 96 and 112% at the fortification level of 0.5 ppb in a river water sample, and their relative standard deviations were between 1 and 15%. The detection limits of these compounds ranged between 0.05 and 0.14 ppb.