EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Dibasic inhibitors of human mast cell tryptase. Part 1: synthesis and optimization of a novel class of inhibitors.
PMID 11055355
Abstract
The synthesis and optimization of a novel class of reversible and active-site directed dibasic inhibitors of human mast cell tryptase are described. The compounds were shown to be both remarkably potent and selective for tryptase with Ki values for optimized inhibitors in the picomolar range.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
X1202
m-Xylylenediamine, 99%
C8H12N2