EMAIL THIS PAGE TO A FRIEND
Methods in enzymology
Assays for transmembrane movement of sphingolipids.
PMID 11070902
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E9030 Endoglycoceramidase II from Rhodococcus sp., aqueous solution