EMAIL THIS PAGE TO A FRIEND
Organic letters
Parallel synthesis and purification using anthracene-tagged substrates.
PMID 11082021
Abstract
[reaction: see text] A new strategy for the synthesis and purification of synthetic intermediates is described using anthracene-tagged ester substrates in conjunction with an N-benzylmaleimide resin. Anthracene chemical tags permit use of standard solution-phase reaction conditions and reaction-monitoring techniques.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
408018
N-Benzylmaleimide, 99%
C11H9NO2