EMAIL THIS PAGE TO A FRIEND
Organic letters
Highly selective Diels-Alder reactions of dienophiles with 1, 3-cyclohexadiene mediated by Yb(OTf)(3).2H(2)O and ultrahigh pressures.
PMID 11082023
Abstract
[reaction: see text] Ultrahigh pressures and catalytic Yb(OTf)(3).2H(2)O were found to mediate Diels-Alder reactions of various electron-deficient dienophiles with 1,3-cyclohexadiene to produce endo-bicyclo[2.2. 2]oct-2-enes in moderate to excellent yield and selectivity. The proposed total synthesis of hapalindole Q based on bicyclo[2.2. 2]oct-2-ene construction by Diels-Alder reaction and subsequent olefin cleavage is outlined. Preliminary results demonstrating the viability of this strategy are presented.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C100005
1,3-Cyclohexadiene, contains 0.05% BHT as inhibitor, 97%
C6H8