EMAIL THIS PAGE TO A FRIEND
Biomaterials
Native and DPPA cross-linked collagen sponges seeded with fetal bovine epiphyseal chondrocytes used for cartilage tissue engineering.
PMID 11085378
Abstract
Collagen-based biomaterials in the form of sponges (bovine type I collagen, both native and cross-linked by treatment with diphenylphosphorylazide, noted control and DPPA sponges respectively) were tested as three-dimensional scaffolds to support chondrocyte proliferation with maintenance of the phenotype in order to form neocartilage. Control and DPPA sponges were initially seeded with 10(6) or 10(7) foetal bovine epiphyseal chondrocytes and maintained for 4 weeks in culture under static conditions in RPMI/NCTC medium with 10% FCS and without addition of fresh ascorbic acid. Both supports were always present during the study and a partial decrease in size and weight was detected only with control sponges, both seeded and unseeded. Cell proliferation was only noted in the 10(6) cells-seeded sponges (4-fold increase after 4 weeks of culture). Specific cartilage collagens (types II and XI) were deposited in the matrix throughout the culture and traces of type I collagen were noticed only in the culture medium after 2-3 weeks and 4 weeks in the case of 10(6) and 10(7) cells-seeded sponges, respectively. Glycosaminoglycans accumulated in the matrix, up to 1.8 and 9.8% of total dry weight after one month with both seeding conditions, which was much lower than in the natural tissue. In the 10(7) cells-seeded sponges, mineral deposition, observed with unseeded sponges, was significantly decreased (2- to 3-fold). These in vitro results indicate that both collagen matrices can support the development of tissue engineered cartilage.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
178756
Diphenyl phosphoryl azide, 97%
C12H10N3O3P