EMAIL THIS PAGE TO A FRIEND
Veterinary research communications
Chemical characterization of bovine urine with special reference to oestrus.
PMID 11085464
Abstract
Cows' urine was analysed by gas chromatography mass spectrometry (GC-MS). The profiles from preovulatory, ovulatory and postovulatory samples were compared to establish any qualitative and quantitative differences that might have potential value in olfactory communication. Dichloromethane was used as the solvent for extraction of the compounds. Seven different compounds were detected, of which only two were common to all the chromatograms. The chemical profile of oestrous urine was distinguished by the presence of two specific compounds, viz. di-n-propyl phthalate and 1-iodoundecane, that were not found in the other samples. As oestrous urine has been shown to elicit sexual behaviour in cattle, these two characteristic peaks may represent important chemical compounds that elicit signals that allow the bull to detect 'oestrous odours'.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45624
Dipropyl phthalate, PESTANAL®, analytical standard
C14H18O4
290602
Dipropyl phthalate, 98%
C14H18O4