EMAIL THIS PAGE TO A FRIEND
Logopedics, phoniatrics, vocology
Linguistic evaluation of Profet II: a pilot project.
PMID 11086802
Abstract
Profet, a word prediction program, was designed to accelerate the writing process and to minimize the writing effort of persons with motor dysfunction. It has also proved to be beneficial in text construction for persons with linguistic impairment such as dyslexia. With increasing linguistic demands on support for individuals with severe reading and writing difficulties/dyslexia, the need for an improved version of Profet arose. Thus, Profet II was designed. In this study, a procedure for evaluating Profet II has been developed. Results from a single-case evaluation study with a person with dyslexia are presented. The possible implications for support and aspects such as spelling, morphology and subjective judgements of and attitudes towards texts are discussed.