EMAIL THIS PAGE TO A FRIEND
Organic letters
Synthesis of 6-acetamido-5-amino- and -5-guanidino-3, 4-dehydro-N-(2-ethylbutyryl)- 3-piperidinecarboxylic acids related to zanamivir and oseltamivir, inhibitors of influenza virus neuraminidases.
PMID 11101432
Abstract
[reaction: see text] 6-Acetamido-5-amino- and -5-guanidino-3, 4-dehydro-N-(2-ethylbutyryl)-3-piperidinecarboxylic acids (8 and 9) have been synthesized starting from natural siastatin B, a bacterial neuraminidase inhibitor isolated from Streptomyces culture in a stereospecific fashion. These compounds are related to zanamivir and oseltamivir, inhibitors of influenza virus neuraminidases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S8063
Siastatin B, lyophilized powder, from microbial
C8H14N2O5