EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
A simple method of preparing D-(minus)-arginine monohydrochloride from DL-arginine.
PMID 1111076
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A6757
D-Arginine monohydrochloride, ≥98% (TLC)
C6H14N4O2 · HCl