EMAIL THIS PAGE TO A FRIEND
American journal of respiratory and critical care medicine
Talc should be used for pleurodesis.
PMID 11112103
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
420433
Talc, −350 mesh
H2Mg3O12Si4