EMAIL THIS PAGE TO A FRIEND
Neurology
Oxidative stress in patients with Friedreich ataxia.
PMID 11113228
Abstract
Increased generation of reactive oxygen species may underlie the pathophysiology of Friedreich ataxia (FRDA). The authors measured concentrations of 8-hydroxy-2'-deoxyguanosine (8OH2'dG), a marker of oxidative DNA damage, in urine and of dihydroxybenzoic acid (DHBA), a marker of hydroxyl radical attack, in plasma of 33 patients with FRDA. They found a 2.6-fold increase in normalized urinary 8OH2'dG but no change in plasma DHBA as compared with controls. Oral treatment with 5 mg/kg/day of the antioxidant idebenone for 8 weeks significantly decreased urinary 8OH2'dG concentrations, indicating that 8OH2'dG may be useful in monitoring therapeutic interventions in patients with FRDA.-1721