EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
An NMR study of the lactonization of alpha-N-acetylneuraminyl-(2 --> 3)-lactose.
PMID 11117332
Abstract
The composition of the products formed by treatment of commercial alpha-Neu5Ac-(2 --> 3)-beta-D-Galp-(1 --> 4)-D-Glc (3'-sialyllactose) with glacial acetic acid was investigated by 1H-13C one- and two-dimensional NMR spectroscopy and fast atom bombardment-mass spectrometry. The data confirmed that the major product of the reaction was alpha-Neu5Ac-(2 --> 3)-beta-D-Galp-(1 --> 4)-D-Glc-(1c --> 2b)-lactone, which reverted to the starting material on standing in aqueous solution at ambient temperature, but for which complete NMR assignments are reported. The NMR data led to the tentative conclusion that the reaction also yielded small amounts of lactose, and alpha-Neu5Ac-(2 --> 3)-beta-D-Galp-(1 --> 4)-D-Glc-(1c --> 4b)-lactone which was stable in aqueous solution.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A8681
3′-Sialyllactose, from bovine milk or colostrum, ≥97% (HPLC)
C23H39NO19