EMAIL THIS PAGE TO A FRIEND
Cancer research
Sulindac sulfone inhibits K-ras-dependent cyclooxygenase-2 expression in human colon cancer cells.
PMID 11118042
Abstract
Both the sulfide and sulfone metabolites of sulindac, a nonsteroidal anti-inflammatory drug, display anticarcinogenic effects in experimental models. Sulindac sulfide inhibits cyclooxygenase (COX) enzyme activities and has been reported to suppress ras-dependent signaling. However, the mechanisms by which sulindac sulfone suppresses cancer growth are not as defined. We studied the effects of these sulindac metabolites in human colon cancer-derived Caco-2 cells that have been transfected with an activated K-ras oncogene. Stable transfected clones expressed high levels of COX-2 mRNA and protein, compared with parental cells. K-ras-transfected cells formed tumors more quickly when injected into severe combined immunodeficiency disease mice than parental cells, and this tumorigenesis was suppressed by treatment with sulindac. Sulindac sulfone inhibited COX-2 protein expression, which resulted in a decrease in prostaglandin synthase E2 production. Sulindac sulfide had little effect on COX-2 in this model, but did suppress prostaglandin synthase E2 production, presumably by inhibiting COX enzyme activity. These data indicate that the sulfide and sulfone derivatives of sulindac exert COX-dependent effects by distinct mechanisms.
Related Materials