EMAIL THIS PAGE TO A FRIEND
Forensic science international
Blood carbofuran concentrations in suicidal ingestion cases.
PMID 11118755
Abstract
We describe four fatal cases due to ingestion of carbofuran, a carbamate insecticide. Carbofuran was detected in the gastric contents using thin layer chromatography (TLC) and gas chromatography/mass spectrophotometry (GC/MS), and quantified in the blood using a gas chromatograph equipped with nitrogen-phosphorus detector (NPD). Fatal concentrations of carbofuran in blood ranged from 0.32 to 11.6 microg/ml.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31544
Benfuracarb, PESTANAL®, analytical standard
C20H30N2O5S