EMAIL THIS PAGE TO A FRIEND
Journal of virology
The ability of integrin alpha(v)beta(3) To function as a receptor for foot-and-mouth disease virus is not dependent on the presence of complete subunit cytoplasmic domains.
PMID 11119622