EMAIL THIS PAGE TO A FRIEND
The Journal of investigative dermatology
Xeroderma pigmentosum variant heterozygotes show reduced levels of recovery of replicative DNA synthesis in the presence of caffeine after ultraviolet irradiation.
PMID 11121129
Abstract
Patients with xeroderma pigmentosum variant show clinical photosensitivity, skin neoplasias induced by ultraviolet light, and defective postreplication repair, but normal nucleotide excision repair. We recently reported an alternative, simple method for the diagnosis of xeroderma pigmentosum variant that measures by autoradiography three cellular markers for DNA repair after ultraviolet irradiation: unscheduled DNA synthesis, recovery of RNA synthesis, and recovery of replicative DNA synthesis. Among hereditary photosensitive disorders, including other xeroderma pigmentosum groups, Cockayne syndrome, and a newly established ultraviolet-sensitive syndrome, only xeroderma pigmentosum variant cells exhibited normal unscheduled DNA synthesis, normal recovery of RNA synthesis, but reduced recovery of replicative DNA synthesis (51 +/- 6% the rate relative to normal controls). This reduction of recovery of replicative DNA synthesis was enhanced in the presence of a nontoxic level of caffeine to 36 +/- 5%. In this study we assess the cellular markers in two independent families that included two photosensitive patients that were identified as xeroderma pigmentosum variant. Cells from heterozygotic parents showed normal levels of unscheduled DNA synthesis, recovery of RNA synthesis, and recovery of replicative DNA synthesis, but reduced rates of recovery of replicative DNA synthesis in the presence of 1 mM caffeine (53 +/- 8% relative to the normal control). Furthermore, with a colony-forming assay, the cells showed normal survival by ultraviolet without caffeine, but slightly reduced survival by ultraviolet with 1 mM caffeine present. In one family, we confirmed inheritance of two heterozygous mis-sense mutations. One mutation is an A-->G transition at nucleotide 1840 that generates a K535E mis-sense mutation. Another mutation is an A-->C transversion at nucleotide 2003 that generates a K589 mis-sense mutation. Each of these mutations were absent in 52 unrelated Japanese individuals. These results suggest that xeroderma pigmentosum variant heterozygotes can be identified by their sensitivity to ultraviolet irradiation in the presence of nontoxic levels of caffeine.