EMAIL THIS PAGE TO A FRIEND
Die Pharmazie
[E-Z ratio and stability of halogenated benzylidene derivatives formed during the detection of glycine conjugates].
PMID 11125998
Abstract
The orange fluorescing spot formed during the detection of halogenated hippuric acids with 4-N,N-Dimethylaminobenzaldehyde on TLC-plates consists of a E/Z-mixture of the corresponding benzyliden derivatives. Intermediates are the azlactones.