EMAIL THIS PAGE TO A FRIEND
Acta poloniae pharmaceutica
Enzymatic hydrolysis of O-acyl esters of oxprenolol.
PMID 11126614
Abstract
The stability of each O-acyl derivative was investigated in a solution of hydrochloric acid, artificial gastric juice, phosphate buffer at pH 7.4 and in artificial intestinal juice. A study of the relationship between the structure and enzymatic hydrolysis of the homologous series of oxprenolol esters was made.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O0250000 Oxprenolol hydrochloride, European Pharmacopoeia (EP) Reference Standard
C15H23NO3 · HCl