EMAIL THIS PAGE TO A FRIEND
International journal of pharmaceutics
Cross-linked beta-cyclodextrin microcapsules: preparation and properties.
PMID 11137335
Abstract
Microcapsules were prepared by interfacial cross-linking of beta-cyclodextrins (beta-CD) with terephthaloyl chloride (TC). Batches were prepared from beta-CD solutions in 1 M NaOH, using 5% TC and a 30 min reaction time. Microcapsules were studied with respect to morphology (microscopy), size (laser diffraction technique) and, for selected batches, IR spectroscopy, determination of beta-CD content (polarimetry after alkaline dissolution of microcapsules) and complexing properties, evaluated using p-nitrophenol (pNP) as the guest molecule. Well-formed microcapsules were obtained from 5, 7.5, and 10% beta-CD solutions. The mean size of all batches was in the 10-35 microm range. The IR spectrum showed bands at 1724, 1280 and 731 cm(-1), reflecting the formation of esters. The beta-CD contents were 46, 56-58 or 60-66% for batches prepared from 5, 7.5 or 10% beta-CD solutions, respectively. The experiments conducted with 1 mM pNP showed a rapid complexation reaching a maximum within 1 h. When incubating 50 mg lyophilized microcapsules in 10 ml pNP solution, the maximal fixation (97.8 micromol/g microcapsules) was observed for small-sized particles ( approximately 11 microm) prepared from a 7.5% beta-CD solution. The method then appears as a simple and rapid procedure to provide stable microcapsules, having an interesting guest-binding ability.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
120871
Terephthaloyl chloride, ≥99%, flakes
C8H4Cl2O2