EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
Pseudoprolines (psiPro) in drug design: direct insertion of psiPro systems into cyclosporin C.
PMID 11140965
Abstract
The insertion of acetals that exhibit variable structural features into complex peptides such as cyclosporin C (CsC) results in oxazolidine derivatives (pseudoprolines, psiPro) of tailored physico-chemical and biological properties. N,O-Acetalation of the 2-threonine hydroxyl group and the preceding amide nitrogen of CsC is achieved by treating the molecule with a number of both arylated and non-arylated dimethyl acetals. The psiPro-containing CsC derivatives exhibit enhanced conformational backbone rigidity, as suggested by analytical HPLC, NMR spectroscopy and by kinetic measurements on binding with their receptor protein cyclophilin A (CypA) that were not time-dependent. IC50 values for calf-thymus CypA were obtained by kinetic evaluation of its cis-->trans isomerase activity. The choice of the para-substituted aryl dimethyl acetals allows the inhibitory properties of the corresponding derivatives to be modulated to either prodrugs or moderately strongly binding cyclosporin C derivatives.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0192 Cyclosporin C, ≥95% (HPLC), from Acremonium luzulae
C62H111N11O13