EMAIL THIS PAGE TO A FRIEND
Anales espanoles de pediatria
[Accidental oral mercurochrome poisoning].
PMID 11141371
Abstract
Neonatal mercury poisoning, especially that due to merbromin ingestion, is uncommon. We describe the case of a 10 day old newborn infant who was given mercurochrome orally for 7 days due to misunderstanding of medical instructions. Initial symptoms included loss of appetite and low weight increase. Elevated blood mercury concentrations were found. Chelating therapy with dimercaprol was initiated and the patient's evolution was good. We discuss the potential toxicity of mercury and emphasise the importance of the transmission of information by physicians, especially to the immigrant population.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M7011
Mercury dibromofluorescein disodium salt, practical grade, crystalline
C20H8Br2HgNa2O6